Product Details

SNP ID
rs77090490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49984863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGGCTAAATGCAGGAATGCAT[A/G]TGGGAAGCACCTGGCAAAGTGCCGG
Phenotype
MIM: 604238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNAI1 PubMed Links

Gene Details

Gene
SNAI1
Gene Name
snail family transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005985.3 Intron NP_005976.2

View Full Product Details