Product Details

SNP ID
rs75218434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58401799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCATATAAAAGGCTGCAATAC[A/G]TTAAATGCAGCATGACCCAAGCTGA
Phenotype
MIM: 605704
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAPB PubMed Links

Gene Details

Gene
VAPB
Gene Name
VAMP associated protein B and C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195677.1 Intron NP_001182606.1
NM_004738.4 Intron NP_004729.1

View Full Product Details