Product Details

SNP ID
rs78017147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36752475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAGACTTGGTCCTAAATGATTT[C/T]TGGATTGTTTCAAAACCTGAAAAAC
Phenotype
MIM: 609175 MIM: 605273
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSN1 PubMed Links

Gene Details

Gene
DSN1
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145315.1 1739 UTR 3 NP_001138787.1
NM_001145316.1 1739 UTR 3 NP_001138788.1
NM_001145317.1 1739 UTR 3 NP_001138789.1
NM_001145318.1 1739 UTR 3 NP_001138790.1
NM_024918.3 1739 UTR 3 NP_079194.3
XM_006723876.2 1739 UTR 3 XP_006723939.1
XM_017028070.1 1739 UTR 3 XP_016883559.1
XM_017028071.1 1739 UTR 3 XP_016883560.1
XM_017028072.1 1739 UTR 3 XP_016883561.1
Gene
NDRG3
Gene Name
NDRG family member 3
There are no transcripts associated with this gene.

View Full Product Details