Product Details

SNP ID
rs79214929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38748902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTGGGTTTGAGGGGAGGGGTG[C/T]GGTCTCGTCTCCGCTCACTCTGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR5 PubMed Links

Gene Details

Gene
ACTR5
Gene Name
ARP5 actin-related protein 5 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024855.3 Intron NP_079131.3

View Full Product Details