Product Details

SNP ID
rs79456487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18383379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGAGTCAAAATTACACATTCTT[A/G]TAAGTGGTTTATTGAGAGCTGATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DZANK1 PubMed Links

Gene Details

Gene
DZANK1
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099407.1 3575 UTR 3 NP_001092877.1
XM_005260742.3 3575 UTR 3 XP_005260799.1
XM_005260743.3 3575 UTR 3 XP_005260800.1
XM_006723584.2 3575 UTR 3 XP_006723647.1
XM_011529268.2 3575 UTR 3 XP_011527570.1
XM_011529271.2 3575 UTR 3 XP_011527573.1
XM_011529272.2 3575 UTR 3 XP_011527574.1
XM_011529275.2 3575 UTR 3 XP_011527577.1
XM_011529277.2 3575 UTR 3 XP_011527579.1
XM_017027909.1 3575 UTR 3 XP_016883398.1
XM_017027910.1 3575 UTR 3 XP_016883399.1
XM_017027911.1 3575 UTR 3 XP_016883400.1
XM_017027912.1 3575 UTR 3 XP_016883401.1
XM_017027913.1 3575 UTR 3 XP_016883402.1
XM_017027914.1 3575 UTR 3 XP_016883403.1
XM_017027915.1 3575 UTR 3 XP_016883404.1
XM_017027916.1 3575 UTR 3 XP_016883405.1
XM_017027917.1 3575 UTR 3 XP_016883406.1
XM_017027918.1 3575 UTR 3 XP_016883407.1
XM_017027919.1 3575 UTR 3 XP_016883408.1
XM_017027920.1 3575 UTR 3 XP_016883409.1
XM_017027921.1 3575 UTR 3 XP_016883410.1
Gene
LINC00851
Gene Name
long intergenic non-protein coding RNA 851
There are no transcripts associated with this gene.

View Full Product Details