Product Details

SNP ID
rs75186990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3163587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGTCTGACCTCAGAAACTGGGC[A/G]AGGAGAGTGCCTGAGAGGCAGGTAG
Phenotype
MIM: 614272 MIM: 610484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASTKD5 PubMed Links

Gene Details

Gene
FASTKD5
Gene Name
FAST kinase domains 5
There are no transcripts associated with this gene.

Gene
LZTS3
Gene Name
leucine zipper tumor suppressor family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282533.1 4251 UTR 3 NP_001269462.1
XM_005260890.3 4251 UTR 3 XP_005260947.1
XM_005260892.1 4251 UTR 3 XP_005260949.1
XM_005260893.2 4251 UTR 3 XP_005260950.1
XM_011529407.2 4251 UTR 3 XP_011527709.1
XM_011529408.2 4251 UTR 3 XP_011527710.1
XM_011529409.2 4251 UTR 3 XP_011527711.1
Gene
UBOX5
Gene Name
U-box domain containing 5
There are no transcripts associated with this gene.

View Full Product Details