Product Details

SNP ID
rs79285051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3735108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATGACTAGAATGCGATTCTCA[A/G]TAAGACCAAAATATCTAACAAGAAA
Phenotype
MIM: 610702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA12B PubMed Links

Gene Details

Gene
HSPA12B
Gene Name
heat shock protein family A (Hsp70) member 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197327.1 Intron NP_001184256.1
NM_001318322.1 Intron NP_001305251.1
NM_052970.4 Intron NP_443202.3
XM_011529151.2 Intron XP_011527453.1
XM_017027632.1 Intron XP_016883121.1
XM_017027633.1 Intron XP_016883122.1
XM_017027634.1 Intron XP_016883123.1
XM_017027635.1 Intron XP_016883124.1

View Full Product Details