Product Details
- SNP ID
-
rs75442118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:145508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCATGTAAAGAATGGTTGGGCAA[C/T]GTGCGGCAAACAAAGGGACTGCTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB126
PubMed Links
Gene Details
- Gene
- DEFB126
- Gene Name
- defensin beta 126
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030931.3 |
228 |
Missense Mutation |
ACG,ATG |
T51M |
NP_112193.1 |
View Full Product Details