Product Details

SNP ID
rs74379631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29194681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTATTCTTTCTTTACCAGGGTGAT[G/T]TTGGTGCATCTGACACCTTTTCAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details