Product Details

SNP ID
rs79824453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32732691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGCATTTAGCCTCCAATTCC[A/T]TTTCTCCCTTGGCTGAGATCTGTTC
Phenotype
MIM: 604297
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PAXBP1 PubMed Links

Gene Details

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
There are no transcripts associated with this gene.

Gene
PAXBP1-AS1
Gene Name
PAXBP1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SYNJ1
Gene Name
synaptojanin 1
There are no transcripts associated with this gene.

View Full Product Details