Product Details

SNP ID
rs78335539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34788103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTGGCTGAAGACACCAGCTTG[A/G]CAGTTCCCCTTTAAGAAGCATTCCA
Phenotype
MIM: 151385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01426 PubMed Links

Gene Details

Gene
LINC01426
Gene Name
long intergenic non-protein coding RNA 1426
There are no transcripts associated with this gene.

Gene
RUNX1
Gene Name
runt related transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001890.2 5470 UTR 3 NP_001001890.1
NM_001122607.1 5470 Intron NP_001116079.1
NM_001754.4 5470 UTR 3 NP_001745.2
XM_005261068.3 5470 UTR 3 XP_005261125.1
XM_005261069.4 5470 UTR 3 XP_005261126.1
XM_011529766.2 5470 UTR 3 XP_011528068.1
XM_011529767.2 5470 UTR 3 XP_011528069.1
XM_011529768.2 5470 UTR 3 XP_011528070.1
XM_011529770.2 5470 Intron XP_011528072.1
XM_017028487.1 5470 UTR 3 XP_016883976.1

View Full Product Details