Product Details

SNP ID
rs77315286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41177195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAATGGAGGCTTGTTCCAGAAT[G/T]CAGAAATCCCTGTTAAATTAGGCAG
Phenotype
MIM: 605668 MIM: 613770
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BACE2 PubMed Links

Gene Details

Gene
BACE2
Gene Name
beta-site APP-cleaving enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012105.4 8045 Intron NP_036237.2
NM_138991.2 8045 Intron NP_620476.1
NM_138992.2 8045 Intron NP_620477.1
XM_017028314.1 8045 Intron XP_016883803.1
Gene
MIR3197
Gene Name
microRNA 3197
There are no transcripts associated with this gene.

Gene
PLAC4
Gene Name
placenta specific 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182832.2 8045 UTR 3 NP_878252.2

View Full Product Details