Product Details

SNP ID
rs76631814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17521022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAGACATTGGTGATACAGACT[G/T]GAGAATTCACTGCTGTTAGGTGGCG
Phenotype
MIM: 602621
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXADR PubMed Links

Gene Details

Gene
CXADR
Gene Name
coxsackie virus and adenovirus receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207063.1 Intron NP_001193992.1
NM_001207064.1 Intron NP_001193993.1
NM_001207065.1 Intron NP_001193994.1
NM_001207066.1 Intron NP_001193995.1
NM_001338.4 Intron NP_001329.1
XM_011529476.2 Intron XP_011527778.1
XM_011529477.2 Intron XP_011527779.1
XM_011529478.2 Intron XP_011527780.1
XM_011529479.1 Intron XP_011527781.1

View Full Product Details