Product Details

SNP ID
rs75593609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41714058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAACTCTCCAGGTCAGGTGACT[A/G]CATCCAGCTGAGACTCCTGGCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00112 PubMed Links

Gene Details

Gene
LINC00112
Gene Name
long intergenic non-protein coding RNA 112
There are no transcripts associated with this gene.

Gene
LINC00479
Gene Name
long intergenic non-protein coding RNA 479
There are no transcripts associated with this gene.

View Full Product Details