Product Details

SNP ID
rs74721445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41680937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGCACAGATGGCTGAGCCTTG[A/C]CCTAGATTCCGTGGGTCTGGAGAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00111 PubMed Links

Gene Details

Gene
LINC00111
Gene Name
long intergenic non-protein coding RNA 111
There are no transcripts associated with this gene.

View Full Product Details