Product Details

SNP ID
rs76214858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36996991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGGAATTGAAAATGAATCCCT[A/G]CTGGGGAAAGATTTCTGGTGTTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX33 PubMed Links

Gene Details

Gene
TEX33
Gene Name
testis expressed 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163857.1 Intron NP_001157329.1
NM_178552.3 Intron NP_848647.1
XM_011530165.2 Intron XP_011528467.1
XM_011530166.1 Intron XP_011528468.1

View Full Product Details