Product Details

SNP ID
rs75119577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20427797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAAGGCAAGTGTGTGCATCCAG[A/G]GAGCTGGAAAAGGTTGGGCTGACGT
Phenotype
MIM: 613619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCARF2 PubMed Links

Gene Details

Gene
SCARF2
Gene Name
scavenger receptor class F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153334.6 Intron NP_699165.3
NM_182895.4 Intron NP_878315.2
XM_017029065.1 Intron XP_016884554.1

View Full Product Details