Product Details

SNP ID
rs80336397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40058760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCCCAGTACTTGAATCTTGAA[A/G]TTCTGACACATTGAATCTGGAAACA
Phenotype
MIM: 610740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNRC6B PubMed Links

Gene Details

Gene
TNRC6B
Gene Name
trinucleotide repeat containing 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024843.1 Intron NP_001020014.1
NM_001162501.1 Intron NP_001155973.1
NM_015088.2 Intron NP_055903.2

View Full Product Details