Product Details
- SNP ID
-
rs76649502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41246643 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTCTGCAGCAGACTGTGGCGGGCG[A/G]AGGAGCAGGATTCCAGGGCGCTGTT
- Phenotype
-
MIM: 616236
MIM: 602362
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHADL
PubMed Links
Gene Details
- Gene
- CHADL
- Gene Name
- chondroadherin like
There are no transcripts associated with this gene.
- Gene
- MIR6889
- Gene Name
- microRNA 6889
There are no transcripts associated with this gene.
- Gene
- RANGAP1
- Gene Name
- Ran GTPase activating protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278651.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
NP_001265580.1 |
NM_001317930.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
NP_001304859.1 |
NM_002883.3 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
NP_002874.1 |
XM_005261695.1 |
2156 |
Missense Mutation |
TCC,TTC |
S630F |
XP_005261752.1 |
XM_005261696.1 |
2156 |
Missense Mutation |
TCC,TTC |
S616F |
XP_005261753.1 |
XM_006724289.3 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_006724352.1 |
XM_011530293.1 |
2156 |
Missense Mutation |
TCC,TTC |
S585F |
XP_011528595.1 |
XM_011530294.2 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_011528596.1 |
XM_011530295.2 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_011528597.1 |
XM_011530297.1 |
2156 |
Intron |
|
|
XP_011528599.1 |
XM_017028893.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_016884382.1 |
XM_017028894.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_016884383.1 |
XM_017028895.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_016884384.1 |
XM_017028896.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_016884385.1 |
XM_017028897.1 |
2156 |
Missense Mutation |
TCC,TTC |
S575F |
XP_016884386.1 |
XM_017028898.1 |
2156 |
Missense Mutation |
TCC,TTC |
S542F |
XP_016884387.1 |
XM_017028899.1 |
2156 |
Missense Mutation |
TCC,TTC |
S530F |
XP_016884388.1 |
View Full Product Details