Product Details
- SNP ID
-
rs79622696
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41975133 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTGAAAGGTAAGTTGGAGCCTT[A/G]TATATATGGAAGTCCTACACAGAAC
- Phenotype
-
MIM: 608314
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEPT3
PubMed Links
Gene Details
- Gene
- SEPT3
- Gene Name
- septin 3
View Full Product Details