Product Details

SNP ID
rs77407491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24586169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTGAGCTTGCGGGCAGTGGCC[C/T]GCGTCAGTCGGTTGCCGTTGAGCAG
Phenotype
MIM: 612346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGT1 PubMed Links

Gene Details

Gene
GGT1
Gene Name
gamma-glutamyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288833.1 694 Intron NP_001275762.1
NM_013421.2 694 Intron NP_038265.2
NM_013430.2 694 Intron NP_038347.2
Gene
LRRC75B
Gene Name
leucine rich repeat containing 75B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207644.2 694 Missense Mutation CAG,CGG Q222R NP_997527.2
XM_005261600.2 694 Intron XP_005261657.1

View Full Product Details