Product Details

SNP ID
rs78334860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50684162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGCACACTGGGAAGCAGATAC[A/G]CCCGGGCCTGTTGGATGACTGGGTG
Phenotype
MIM: 606230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHANK3 PubMed Links

Gene Details

Gene
SHANK3
Gene Name
SH3 and multiple ankyrin repeat domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033517.1 Intron NP_277052.1

View Full Product Details