Product Details

SNP ID
rs77252560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135906991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGCCAACCCCAACCTGATAAC[A/T]TCTTGTCAACTGCTTCAACAGGGCT
Phenotype
MIM: 300359
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SAGE1 PubMed Links

Gene Details

Gene
SAGE1
Gene Name
sarcoma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018666.2 977 Missense Mutation ATC,TTC I268F NP_061136.2
XM_017029621.1 977 Missense Mutation ATC,TTC I268F XP_016885110.1
XM_017029622.1 977 Intron XP_016885111.1
XM_017029623.1 977 Missense Mutation ATC,TTC I268F XP_016885112.1
XM_017029624.1 977 Missense Mutation ATC,TTC I237F XP_016885113.1
XM_017029625.1 977 Missense Mutation ATC,TTC I268F XP_016885114.1
XM_017029626.1 977 Missense Mutation ATC,TTC I29F XP_016885115.1
XM_017029627.1 977 Intron XP_016885116.1

View Full Product Details