Product Details
- SNP ID
-
rs76185484
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:123624068 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAATTTTTTTTTTACCTTGGTTA[A/G]TTTGTAATGCCATTTATGTACAACA
- Phenotype
-
MIM: 300395
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
THOC2
PubMed Links
Gene Details
- Gene
- THOC2
- Gene Name
- THO complex 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001081550.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1104L |
NP_001075019.1 |
XM_005262450.3 |
3492 |
Silent Mutation |
CTA,TTA |
L1104L |
XP_005262507.1 |
XM_017029662.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885151.1 |
XM_017029663.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885152.1 |
XM_017029664.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885153.1 |
XM_017029665.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885154.1 |
XM_017029666.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1150L |
XP_016885155.1 |
XM_017029667.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885156.1 |
XM_017029668.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885157.1 |
XM_017029669.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885158.1 |
XM_017029670.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1150L |
XP_016885159.1 |
XM_017029671.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885160.1 |
XM_017029672.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885161.1 |
XM_017029673.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1108L |
XP_016885162.1 |
XM_017029674.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1104L |
XP_016885163.1 |
XM_017029675.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885164.1 |
XM_017029676.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1154L |
XP_016885165.1 |
XM_017029677.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1108L |
XP_016885166.1 |
XM_017029678.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1104L |
XP_016885167.1 |
XM_017029679.1 |
3492 |
Silent Mutation |
CTA,TTA |
L1108L |
XP_016885168.1 |
View Full Product Details