Product Details

SNP ID
rs77400381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19912079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGCGTATGTGACAGAATGTGA[A/G]TCAGGACATAAAAACATTACAAAAC
Phenotype
MIM: 613602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR35 PubMed Links

Gene Details

Gene
WDR35
Gene Name
WD repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006657.1 3825 UTR 3 NP_001006658.1
NM_020779.3 3825 UTR 3 NP_065830.2
XM_011533007.2 3825 UTR 3 XP_011531309.1

View Full Product Details