Product Details

SNP ID
rs74600365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:175638033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCAGGGCTTATTACTGTCCCT[C/T]GCTAGGTGAACAGTGATTACTGAAA
Phenotype
MIM: 601275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPM6A PubMed Links

Gene Details

Gene
GPM6A
Gene Name
glycoprotein M6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261447.1 Intron NP_001248376.1
NM_001261448.1 Intron NP_001248377.1
NM_005277.4 Intron NP_005268.1
NM_201591.2 Intron NP_963885.1
NM_201592.2 Intron NP_963886.1
XM_006714183.1 Intron XP_006714246.1
XM_011531877.1 Intron XP_011530179.1
XM_017008036.1 Intron XP_016863525.1

View Full Product Details