Product Details

SNP ID
rs77782933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184090368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGGGAAGGCAGGCAGGCCATC[A/C]ATTTGTCCATCAATATGCAGGGCCT
Phenotype
MIM: 616983
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENPP6 PubMed Links

Gene Details

Gene
ENPP6
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153343.3 2274 UTR 3 NP_699174.1

View Full Product Details