Product Details

SNP ID
rs74953250
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185162559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTAGAAGATACACATGCATA[C/T]CACTATTTCTCTATTAAGAACACAT
Phenotype
MIM: 616047
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CFAP97 PubMed Links

Gene Details

Gene
CFAP97
Gene Name
cilia and flagella associated protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292033.1 1985 Intron NP_001278962.1
NM_020827.2 1985 UTR 3 NP_065878.1
XM_017008483.1 1985 UTR 3 XP_016863972.1
XM_017008484.1 1985 UTR 3 XP_016863973.1
XM_017008485.1 1985 UTR 3 XP_016863974.1
XM_017008486.1 1985 UTR 3 XP_016863975.1

View Full Product Details