Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042402.1 | 1738 | Intron | NP_001035861.1 | ||
NM_014435.3 | 1738 | UTR 3 | NP_055250.2 | ||
XM_005262920.3 | 1738 | Intron | XP_005262977.1 | ||
XM_005262921.3 | 1738 | Intron | XP_005262978.1 | ||
XM_005262923.2 | 1738 | UTR 3 | XP_005262980.1 | ||
XM_005262924.2 | 1738 | UTR 3 | XP_005262981.1 | ||
XM_005262925.3 | 1738 | Intron | XP_005262982.1 | ||
XM_006714180.2 | 1738 | Intron | XP_006714243.1 | ||
XM_011531852.2 | 1738 | Intron | XP_011530154.1 | ||
XM_011531854.2 | 1738 | Intron | XP_011530156.1 | ||
XM_017008027.1 | 1738 | Intron | XP_016863516.1 | ||
XM_017008028.1 | 1738 | UTR 3 | XP_016863517.1 | ||
XM_017008029.1 | 1738 | Intron | XP_016863518.1 | ||
XM_017008030.1 | 1738 | UTR 3 | XP_016863519.1 |