Product Details

SNP ID
rs75235734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89240273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATCTTAAGGAACTCAAGACAC[A/G]AGGCAATTTAAGTGCCCTGGAACCC
Phenotype
MIM: 611241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPRIN3 PubMed Links

Gene Details

Gene
GPRIN3
Gene Name
GPRIN family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198281.2 10155 Intron NP_938022.2
XM_005262936.3 10155 UTR 3 XP_005262993.1
XM_005262937.4 10155 UTR 3 XP_005262994.1
XM_017008043.1 10155 UTR 3 XP_016863532.1
XM_017008044.1 10155 UTR 3 XP_016863533.1

View Full Product Details