Product Details
- SNP ID
-
rs77244248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:89888195 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTGTAGTTTCTCTTTAGCGCAA[C/T]GGGTTTCTCTAGTATATCTTATAGG
- Phenotype
-
MIM: 601456
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MMRN1
PubMed Links
Gene Details
- Gene
- MMRN1
- Gene Name
- multimerin 1
View Full Product Details