Product Details

SNP ID
rs80289050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:94301327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACTAGGTGGTAGTGTGCCTAGT[A/G]TCTGGTTGTCTAACTTTTTTGTGAT
Phenotype
MIM: 602598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPGDS PubMed Links

Gene Details

Gene
HPGDS
Gene Name
hematopoietic prostaglandin D synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014485.2 Intron NP_055300.1
XM_005262932.2 Intron XP_005262989.1

View Full Product Details