Product Details

SNP ID
rs79147628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98276339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGACACAAACATTCCATCTAT[A/G]ACAAGTACTCTTTCTAAAATGTAAA
Phenotype
MIM: 179502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAP1GDS1 PubMed Links

Gene Details

Gene
RAP1GDS1
Gene Name
Rap1 GTPase-GDP dissociation stimulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100426.1 Intron NP_001093896.1
NM_001100427.1 Intron NP_001093897.1
NM_001100428.1 Intron NP_001093898.1
NM_001100429.1 Intron NP_001093899.1
NM_001100430.1 Intron NP_001093900.1
NM_021159.4 Intron NP_066982.3

View Full Product Details