Product Details

SNP ID
rs79449940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106058456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTTCTATGAGGATTTAAGAGT[G/T]ATACTTAGTTCCAGACTTGAGTTGT
Phenotype
MIM: 616899
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TBCK PubMed Links

Gene Details

Gene
TBCK
Gene Name
TBC1 domain containing kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163435.2 Intron NP_001156907.1
NM_001163436.2 Intron NP_001156908.1
NM_001163437.2 Intron NP_001156909.1
NM_001290768.1 Intron NP_001277697.1
NM_033115.4 Intron NP_149106.2
XM_011532417.2 Intron XP_011530719.1
XM_017008846.1 Intron XP_016864335.1
XM_017008847.1 Intron XP_016864336.1
XM_017008848.1 Intron XP_016864337.1
XM_017008849.1 Intron XP_016864338.1

View Full Product Details