Product Details

SNP ID
rs77478927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107932965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTATGACCTTGGGCCTGGATGT[A/C]AGTCTTCACCACTGTTCTGACATCT
Phenotype
MIM: 610670
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP2U1 PubMed Links

Gene Details

Gene
CYP2U1
Gene Name
cytochrome P450 family 2 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183075.2 Intron NP_898898.1
XM_005262717.2 Intron XP_005262774.1
XM_005262720.2 Intron XP_005262777.1
Gene
LOC101929595
Gene Name
uncharacterized LOC101929595
There are no transcripts associated with this gene.

View Full Product Details