Product Details

SNP ID
rs80020440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112147911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTTTTCTACTTGGCACGCTCAC[C/T]TCGTGTTCCGGAGTCATGGATAGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf32 PubMed Links

Gene Details

Gene
C4orf32
Gene Name
chromosome 4 open reading frame 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152400.2 Intron NP_689613.1
XM_017007732.1 Intron XP_016863221.1

View Full Product Details