Product Details

SNP ID
rs74956793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112276683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTACTGCCCAGGAATAGACTGAC[A/G]GTCTGAATCAAGATGAGGTTTAAAG
Phenotype
MIM: 607347 MIM: 610851 MIM: 609028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALPK1 PubMed Links

Gene Details

Gene
ALPK1
Gene Name
alpha kinase 1
There are no transcripts associated with this gene.

Gene
AP1AR
Gene Name
adaptor related protein complex 1 associated regulatory protein
There are no transcripts associated with this gene.

Gene
TIFA
Gene Name
TRAF interacting protein with forkhead associated domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052864.2 2016 UTR 3 NP_443096.1

View Full Product Details