Product Details
- SNP ID
-
rs75351777
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112310922 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGAACTTTGGGAGAGCGGAAGTA[A/G]CACTACAGAATCATAGAATCGAAAG
- Phenotype
-
MIM: 607347
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALPK1
PubMed Links
Gene Details
- Gene
- ALPK1
- Gene Name
- alpha kinase 1
View Full Product Details