Product Details
- SNP ID
-
rs76857923
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112708160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAATAGAAAAACGGAAGGGAAAAC[A/C]GTGTTGTTCTACTAAGTGGTGAAAT
- Phenotype
-
MIM: 106410
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANK2
PubMed Links
Gene Details
- Gene
- ANK2
- Gene Name
- ankyrin 2, neuronal
View Full Product Details