Product Details

SNP ID
rs76092860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122925414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCTTTCTGGGCTCCCACTGCT[A/G]TCAGAGTAATGACCAGCATCTTGAT
Phenotype
MIM: 606261 MIM: 613940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT6 PubMed Links

Gene Details

Gene
NUDT6
Gene Name
nudix hydrolase 6
There are no transcripts associated with this gene.

Gene
SPATA5
Gene Name
spermatogenesis associated 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317799.1 Intron NP_001304728.1
NM_145207.2 Intron NP_660208.2
XM_005262783.4 Intron XP_005262840.1
XM_011531678.2 Intron XP_011529980.1
XM_011531679.2 Intron XP_011529981.1
XM_017007825.1 Intron XP_016863314.1
XM_017007826.1 Intron XP_016863315.1
XM_017007827.1 Intron XP_016863316.1
XM_017007828.1 Intron XP_016863317.1
XM_017007829.1 Intron XP_016863318.1
XM_017007830.1 Intron XP_016863319.1

View Full Product Details