Product Details

SNP ID
rs75167013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128269752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAATAAAGTAATTGATATATGA[A/G]GGGCTTTTAGTAAGGGCTTTTTCAA
Phenotype
MIM: 607735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGRMC2 PubMed Links

Gene Details

Gene
PGRMC2
Gene Name
progesterone receptor membrane component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006320.4 3275 UTR 3 NP_006311.2
XM_011531533.1 3275 Intron XP_011529835.1

View Full Product Details