Product Details

SNP ID
rs75601177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155765182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAACATTTACTAAGCATTGTTA[C/T]GATTTGCAAGAACAAAGTCCTTTCT
Phenotype
MIM: 139397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY1B3 PubMed Links

Gene Details

Gene
GUCY1B3
Gene Name
guanylate cyclase 1 soluble subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000857.3 Intron NP_000848.1
NM_001291951.1 Intron NP_001278880.1
NM_001291952.1 Intron NP_001278881.1
NM_001291953.1 Intron NP_001278882.1
NM_001291954.1 Intron NP_001278883.1
NM_001291955.1 Intron NP_001278884.1
XM_011531901.1 Intron XP_011530203.1
XM_017008130.1 Intron XP_016863619.1
XM_017008131.1 Intron XP_016863620.1
XM_017008132.1 Intron XP_016863621.1
XM_017008133.1 Intron XP_016863622.1

View Full Product Details