Product Details

SNP ID
rs77525661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82906360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACATCTTAAAGGTAAAGCAAGA[A/C]AAAAAATCCTAAAACAACCTCTTCC
Phenotype
MIM: 610257 MIM: 612537
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC31A PubMed Links

Gene Details

Gene
SEC31A
Gene Name
SEC31 homolog A, COPII coat complex component
There are no transcripts associated with this gene.

Gene
SNORD144
Gene Name
small nucleolar RNA, C/D box 144
There are no transcripts associated with this gene.

Gene
THAP9
Gene Name
THAP domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317776.1 413 UTR 5 NP_001304705.1
NM_024672.5 413 Missense Mutation AAA,CAA K105Q NP_078948.3
XM_005263220.2 413 UTR 5 XP_005263277.1
XM_011532246.2 413 UTR 5 XP_011530548.1
XM_017008603.1 413 Missense Mutation AAA,CAA K118Q XP_016864092.1
XM_017008604.1 413 UTR 5 XP_016864093.1
XM_017008605.1 413 UTR 5 XP_016864094.1
XM_017008606.1 413 UTR 5 XP_016864095.1
XM_017008607.1 413 UTR 5 XP_016864096.1
XM_017008608.1 413 UTR 5 XP_016864097.1
XM_017008609.1 413 UTR 5 XP_016864098.1
XM_017008610.1 413 UTR 5 XP_016864099.1
Gene
THAP9-AS1
Gene Name
THAP9 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details