Product Details

SNP ID
rs75237162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:161384794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGGTGTTAATTTCCCTATATA[G/T]GGTTTACTTTAGCAGACAAAAGTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FSTL5 PubMed Links

Gene Details

Gene
FSTL5
Gene Name
follistatin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128427.2 3930 UTR 3 NP_001121899.1
NM_001128428.2 3930 UTR 3 NP_001121900.1
NM_020116.4 3930 UTR 3 NP_064501.2
XM_011532126.1 3930 UTR 3 XP_011530428.1

View Full Product Details