Product Details

SNP ID
rs80345192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165080064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTGCTTTATTTAAGAAGGATC[C/T]TAATTTGTTAGAGTAAGATAATTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM192 PubMed Links

Gene Details

Gene
TMEM192
Gene Name
transmembrane protein 192
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100389.1 Intron NP_001093859.1
XM_011531717.2 Intron XP_011530019.1
XM_011531718.2 Intron XP_011530020.1

View Full Product Details