Product Details
- SNP ID
-
rs75259705
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:166734925 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGATTTCAACTGTCATCAATCAA[A/T]TGTATACATCATGGTCATCACCACC
- Phenotype
-
MIM: 607989
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPOCK3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1057377] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPOCK3
- Gene Name
- sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040159.1 |
1334 |
Missense Mutation |
AAT,ATT |
N433I |
NP_001035249.1 |
NM_001204352.1 |
1334 |
Missense Mutation |
AAT,ATT |
N385I |
NP_001191281.1 |
NM_001204353.1 |
1334 |
Missense Mutation |
AAT,ATT |
N316I |
NP_001191282.1 |
NM_001204354.1 |
1334 |
Missense Mutation |
AAT,ATT |
N344I |
NP_001191283.1 |
NM_001204355.1 |
1334 |
Missense Mutation |
AAT,ATT |
N340I |
NP_001191284.1 |
NM_001204356.1 |
1334 |
Missense Mutation |
AAT,ATT |
N393I |
NP_001191285.1 |
NM_001204358.1 |
1334 |
Intron |
|
|
NP_001191287.1 |
NM_001251967.1 |
1334 |
Missense Mutation |
AAT,ATT |
N338I |
NP_001238896.1 |
NM_016950.2 |
1334 |
Missense Mutation |
AAT,ATT |
N436I |
NP_058646.2 |
XM_011532018.1 |
1334 |
Missense Mutation |
AAT,ATT |
N436I |
XP_011530320.1 |
XM_017008257.1 |
1334 |
Missense Mutation |
AAT,ATT |
N445I |
XP_016863746.1 |
XM_017008258.1 |
1334 |
Missense Mutation |
AAT,ATT |
N433I |
XP_016863747.1 |
View Full Product Details