Product Details

SNP ID
rs79876001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135080400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAAACCCATTATTTCAGAATAC[A/G]TGGAATTCATTCAGTCTTTGCACCA
Phenotype
MIM: 605366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFM1 PubMed Links

Gene Details

Gene
OLFM1
Gene Name
olfactomedin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282611.1 Intron NP_001269540.1
NM_001282612.1 Intron NP_001269541.1
NM_006334.3 Intron NP_006325.1
NM_014279.4 Intron NP_055094.1

View Full Product Details