Product Details

SNP ID
rs78105956
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:9730220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGTGTCCTCTCCCCGGGGGGA[A/G]ACTCCAGACACAAACGCGGGGCCTG
Phenotype
MIM: 611321 MIM: 602839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLSTN1 PubMed Links

Gene Details

Gene
CLSTN1
Gene Name
calsyntenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009566.2 4027 UTR 3 NP_001009566.1
NM_001302883.1 4027 UTR 3 NP_001289812.1
NM_014944.4 4027 UTR 3 NP_055759.3
Gene
PIK3CD
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
There are no transcripts associated with this gene.

View Full Product Details