Product Details

SNP ID
rs79330208
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:11696101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGTCCTGCCCTGCCATCGATCA[A/C]CCCACTGAGGACAGAGGACGCAGCC
Phenotype
MIM: 612682 MIM: 604094
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DRAXIN PubMed Links

Gene Details

Gene
DRAXIN
Gene Name
dorsal inhibitory axon guidance protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198545.3 Intron NP_940947.3
XM_011541405.2 Intron XP_011539707.1
XM_017001214.1 Intron XP_016856703.1
XM_017001215.1 Intron XP_016856704.1
Gene
MAD2L2
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
There are no transcripts associated with this gene.

View Full Product Details