Product Details

SNP ID
rs75756949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127941806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTTAACAGGGGTGCACCCGAA[A/C]GCAAGAAGGCAGCTCACTATGTCCA
Phenotype
MIM: 609697
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SAP130 PubMed Links

Gene Details

Gene
SAP130
Gene Name
Sin3A associated protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145928.1 3455 UTR 3 NP_001139400.1
NM_024545.3 3455 UTR 3 NP_078821.2
XM_005263767.3 3455 UTR 3 XP_005263824.1
XM_005263768.3 3455 UTR 3 XP_005263825.1
XM_005263769.3 3455 UTR 3 XP_005263826.1
XM_005263770.3 3455 UTR 3 XP_005263827.1
XM_006712748.3 3455 UTR 3 XP_006712811.1
XM_006712749.3 3455 UTR 3 XP_006712812.1
XM_017004903.1 3455 UTR 3 XP_016860392.1
XM_017004904.1 3455 UTR 3 XP_016860393.1
XM_017004905.1 3455 UTR 3 XP_016860394.1

View Full Product Details